Thermo Scientific™ Nalgene™ PMP Plastic Graduated Cylinders
Guarantee the accuracy requirements of ASTM Class B, E1272 when using these cylinders. All sizes have a generous pour spout and molded-in graduations.
$63.79 - $768.18
Specifications
Material | PMP |
---|---|
Certifications/Compliance | ASTM E1272, CFR 21, ISO 6706, Part 177.1520 |
No. per Pack | 1 |
Calibration | To Contain, To Deliver |
Autoclavable | Autoclavable |
Catalog Number | Mfr. No. | Capacity (Metric) | Graduations | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Graduations | Price | Quantity | ||||
08-572-5A
|
Thermo Scientific™
36630010 |
10 mL | 0.2 mL |
Each for $63.79
Case of 24 Each for $1,530.96
|
|
||||
08-572-5B
|
Thermo Scientific™
36630025 |
25 mL | 0.5 mL |
Each for $70.29
Case of 18 Each for $1,265.22
|
|
||||
08-572-5C
|
Thermo Scientific™
36630050 |
50 mL | 1 mL |
Each for $77.41
Case of 18 Each for $1,393.38
|
|
||||
08-572-5D
|
Thermo Scientific™
36630100 |
100 mL | 1 mL |
Each for $86.62
Case of 12 Each for $1,039.44
|
|
||||
08-572-5E
|
Thermo Scientific™
36630250 |
250 mL | 2 mL |
Each for $116.53
Case of 12 Each for $1,398.36
|
|
||||
08-572-5F
|
Thermo Scientific™
36630500 |
500 mL | 5 mL |
Each for $152.82
Case of 8 Each for $1,222.56
|
|
||||
08-572-5G
|
Thermo Scientific™
36631000 |
1000 mL | 10 mL |
Each for $193.29
Case of 6 Each for $1,159.74
|
|
||||
08-572-5H
|
Thermo Scientific™
36632000 |
2000 mL | 20 mL |
Each for $320.71
Case of 4 Each for $1,282.84
|
|
||||
08-572-5J
|
Thermo Scientific™
36634000 |
4000 mL | 50 mL |
Each for $768.18
Case of 2 Each for $1,536.36
|
|
||||
Description
Meet accuracy requirements of ASTM Class B, E1272 and CFR21, Part 177.1520 (except 2L and 4L sizes) by using Thermo Scientific™ Nalgene™ PMP Graduated Cylinders. All sizes have a generous pour spout, molded-in graduations that won't rub off and a large blue base to prevent tipping. These polymethylpentene cylinders can be chemically sterilized and are suitable for food and beverage use.
Features:- No meniscus to confuse readings—eliminates guesswork
- Large blue polypropylene octagonal base prevents rolling and tipping
- Molded-in graduations
- Excellent chemical resistance for a wide variety of solutions
- Resists temperatures up to 135°C (275°F) for a variety of uses
- Autoclavable (will affect accuracy)
- Transparent
Ordering Information:
- Individually packaged
- 10, 25, 50, 100, 250, 500 and 1000mL sizes have front-to-back raised graduations
- 2000 and 4000mL sizes have standard graduations
Specifications
PMP | |
1 | |
Autoclavable | |
Round | |
Hexagonal |
ASTM E1272, CFR 21, ISO 6706, Part 177.1520 | |
To Contain, To Deliver | |
145°C | |
293°F | |
Graduated Cylinder |