missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Nalgene™ 50mm Inline Syringe Filter with PTFE Membrane

Filter aggressive chemicals, including acids and non-aqueous solvents such as those used in GC and HPLC with this in-line syringe filter with PTFE membrane.

$459.06 - $465.41

Specifications

Diameter (Metric) Outer 50mm
No. per Case 10
No. per Pack 10
View More Specs

Products 2
Catalog Number Mfr. No. Pore Size Price Quantity  
Catalog Number Mfr. No. Pore Size Price Quantity  
09-740-37K View Documents Thermo Scientific™
NDS02220020
0.22μm Case of 10 Each for $465.41
09-740-37L View Documents Thermo Scientific™
NDS02220045
0.45μm Case of 10 Each for $459.06
Description

Description

Filter aggressive chemicals, including acids and non-aqueous solvents, such as those used in GC and HPLC with the Thermo Scientific™ Nalgene™ 50mm Inline Syringe Filter with PTFE membrane. Also for sterile filtration of air and gases. Provides an excellent barrier for sterile venting of benchtop fermentors, or protecting vacuum pumps from moisture. Autoclavable.

Useful for benchtop fermenters, in-line gas filtration for sterile air or CO2 and to protect vacuum pumps from damage by aqueous solutions. Also designed for filtration of aggressive chemicals and solvents such as those used in GC and HPLC.
  • Sample volume size: 0.2 to 5.0L
  • Inlet/Outlet: Stepped hose barb accepts 0.25 to 0.375in. (6 to 9mm) I.D. tubing
  • 0.2 and 0.45μm pore sizes
  • Autoclavable
  • Polypropylene housing
Warranty: 90 days
Specifications

Specifications

50mm
10
10

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.