missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

n-Propyl Alcohol, 99.5% min.,OmniSolv™, MilliporeSigma™

Supplier:  Technidata PX18246

Catalog No. MPX18246


Description

Description

Specifications

Specifications

n-Propyl Alcohol
Colorless
Liquid
C3H8O
1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol
CCCO
60.10
CHEBI:28831
71-23-8
1 L
99.5% min.
MFCD00002941
BDERNNFJNOPAEC-UHFFFAOYSA-N
propan-1-ol
1031
60.1

Chemical Identifiers

71-23-8
60.10
BDERNNFJNOPAEC-UHFFFAOYSA-N
1031
propan-1-ol
C3H8O
MFCD00002941
1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol
CHEBI:28831
CCCO
SDS
Documents

Documents

Promotions

Promotions