Learn More
n-Pentane, Spectrophotometric Grade, 99+% min., Thermo Scientific Chemicals
$129.67 - $828.26
Chemical Identifiers
CAS | 109-66-0 |
---|---|
Molecular Formula | C5H12 |
Molecular Weight (g/mol) | 72.15 |
MDL Number | MFCD00009498 |
InChI Key | OFBQJSOFQDEBGM-UHFFFAOYSA-N |
Synonym | n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish |
PubChem CID | 8003 |
ChEBI | CHEBI:37830 |
IUPAC Name | pentane |
SMILES | CCCCC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA32449K2
|
Thermo Scientific Chemicals
032449K2 |
1 L |
Each for $129.67
|
|
|||||
AA32449M1
|
Thermo Scientific Chemicals
032449M1 |
4 x 1 L |
Each for $313.71
|
|
|||||
AA32449K7
|
Thermo Scientific Chemicals
032449K7 |
4 L |
Each for $286.30
|
|
|||||
AA32449M6
|
Thermo Scientific Chemicals
032449M6 |
4 x 4 L |
Each for $828.26
|
|
|||||
Description
n-Pentane is used in spectrophotometry and environmental testing. It is used in consumer merchandise such as spot lifters/cleaners and foaming shave gels and in commercial products such as a blowing agent for expanded polystyrene (foam) insulation. It may also be used as a solvent (in pure form) in laboratory, chemical analysis, and other settings where a highly volatile non-polar solvent is needed.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
109-66-0 | |
72.15 | |
OFBQJSOFQDEBGM-UHFFFAOYSA-N | |
8003 | |
pentane |
C5H12 | |
MFCD00009498 | |
n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish | |
CHEBI:37830 | |
CCCCC |
Specifications
°C to 130°C | |
Colorless | |
0.626 | |
1 L | |
Liquid | |
>99% | |
MFCD00009498 | |
n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish | |
OFBQJSOFQDEBGM-UHFFFAOYSA-N | |
pentane | |
8003 | |
72.15 | |
n-Pentane |
109-66-0 | |
Colorless liquid | |
36°C | |
-49°C (-57°F) | |
1.358 | |
C5H12 | |
554 hPa (416mm Hg) at 20°C | |
Miscible with water. | |
CCCCC | |
72.15 | |
CHEBI:37830 | |
Spectrophotometric |
Safety and Handling
P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P301+P310-P303+P361+P353-P304+P340-P312-P331-P370+P378q-P501c
H225-H304-H336
DOTInformation : DOT Class: 3; Packing Group: II
EINECSNumber : 203-692-4
RTECSNumber : RZ9450000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only