missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

n-Pentane, 99+%, for residue analysis, ECD tested for pesticide analysis, Thermo Scientific Chemicals

$160.34 - $160.34

Chemical Identifiers

CAS 109-66-0
Molecular Formula C5H12
Molecular Weight (g/mol) 72.15
MDL Number MFCD00009498
InChI Key OFBQJSOFQDEBGM-UHFFFAOYSA-N
Synonym n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish
PubChem CID 8003
ChEBI CHEBI:37830
IUPAC Name pentane
SMILES CCCCC
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC326640010
SDS View Documents Promotion Details
Thermo Scientific Chemicals
326640010
1 L Glass Bottle
Each for $160.34
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For residue analysis
Specifications

Chemical Identifiers

109-66-0
72.15
OFBQJSOFQDEBGM-UHFFFAOYSA-N
8003
pentane
C5H12
MFCD00009498
n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish
CHEBI:37830
CCCCC

Specifications

-130.0°C
99.0
0.6260g/mL
1 L
Glass Bottle
1.358
99+%
C5H12
MFCD00009498
0.005% max. (Coulometric)
15, 7228
Solubility in water: insoluble. Other solubilities: soluble in alcohol and most other common organic, solvents
CCCCC
72.15
CHEBI:37830
≥99%
n-Pentane, ECD Tested, for Pesticide Analysis
109-66-0
100.0
36.0°C
−49°C
Liquid
0.0005% max.
0.626
CH3(CH2)3CH3
0.25 mPa.s (20°C)
01, 130
n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish
OFBQJSOFQDEBGM-UHFFFAOYSA-N
pentane
8003
72.15
Residue Analysis
Safety and Handling

Safety and Handling

GHS P Statement May be fatal if swallowed and enters airways. May cause drowsiness or dizziness. Toxic to aquatic life with long lasting effects. Highly flammable liquid and vapor. Repeated exposure may cause skin dryness or cracking.

GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician. Do NOT induce vomiting. Avoid release to the environment. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Ground/Bond container and receiving equipment.

Danger

EINECSNumber : 203-692-4

RTECSNumber : RZ9450000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only