missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

n-Pentane, +99%, extra pure, ACROS Organics™

$86.13 - $1,387.15

Chemical Identifiers

CAS 109-66-0
Molecular Formula C5H12
Molecular Weight (g/mol) 72.151
MDL Number MFCD00009498
InChI Key OFBQJSOFQDEBGM-UHFFFAOYSA-N
Synonym n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish
PubChem CID 8003
ChEBI CHEBI:37830
IUPAC Name pentane
SMILES CCCCC
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC170070025 SDS Accessories View Documents Acros Organics
170070025
2.5L Glass Bottle Each for $200.79
AC170070010 SDS View Documents Acros Organics
170070010
1L Glass Bottle Each for $86.13
AC170070250 SDS View Documents Acros Organics
170070250
25L Metal drum Each for $1,387.15
AC170070050 SDS View Documents Acros Organics
AC170070050
5L Metal drum Each for $351.11
Specifications

Chemical Identifiers

109-66-0
72.151
OFBQJSOFQDEBGM-UHFFFAOYSA-N
8003
pentane
C5H12
MFCD00009498
n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish
CHEBI:37830
CCCCC

Specifications

109-66-0
100.0
99+%
CH3(CH2)3CH3
01, 130
n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish
CCCCC
72.151
CHEBI:37830
Liquid
Extra Pure
Glass Bottle
0.005% max.
0.626
−49°C
36.0°C
-130.0°C
n-Pentane
99.0
Vapor Pressure: 573mbar at 20°C
C5H12
MFCD00009498
15, 7228
OFBQJSOFQDEBGM-UHFFFAOYSA-N
pentane
8003
72.15
≥99%
0.6260g/mL
1.358
Solubility in water: insoluble. Other solubilities: soluble in alcohol and most other common organic, solvents
0.05% max. (K.F.)
0.25 mPa.s (20°C)
Undesignated
2.5L
Determine assay (GC) instead of accepting value from supplier's Certificate of Analysi
Safety and Handling

Safety and Handling

GHS P Statement May be fatal if swallowed and enters airways. May cause drowsiness or dizziness. Toxic to aquatic life with long lasting effects. Highly flammable liquid and vapor. Repeated exposure may cause skin dryness or cracking.

GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician. Do NOT induce vomiting. Avoid release to the environment. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Ground/Bond container and receiving equipment.

Danger