Learn More
N,N-Dimethylformamide, 99.8%, Extra Dry over Molecular Sieve, AcroSeal™, Thermo Scientific Chemicals
N,N-Dimethylformamide, 99.8%, C3H7NO, CAS Number-68-12-2, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dwumetyloformamid, dimethylformamide, dmfa, dimethyl formamide, dimetilformamide, dmf, dimethylformamid, 1L, 0.01 % max., Vapor Pressure: 4.9mbar at 20 deg.C, 04, 58
$73.91 - $340.63
Chemical Identifiers
CAS | 68-12-2 |
---|---|
Molecular Formula | C3H7NO |
Molecular Weight (g/mol) | 73.10 |
MDL Number | MFCD00003284 |
InChI Key | ZMXDDKWLCZADIW-UHFFFAOYSA-N |
Synonym | dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa |
PubChem CID | 6228 |
ChEBI | CHEBI:17741 |
IUPAC Name | N,N-dimethylformamide |
SMILES | CN(C)C=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC348431000
|
Thermo Scientific Chemicals
348431000 |
100 mL | AcroSeal™ Glass Bottle |
Each for $73.91
|
|
||||
AC348435000
|
Thermo Scientific Chemicals
348435000 |
500 mL | AcroSeal™ Glass Bottle |
Each for $138.91
|
|
||||
AC348430010
|
Thermo Scientific Chemicals
348430010 |
1 L | AcroSeal™ Glass Bottle |
Each for $205.96
|
|
||||
AC348430025
|
Thermo Scientific Chemicals
348430025 |
2.5 L | AcroSeal™ Glass Bottle |
Each for $340.63
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
68-12-2 | |
73.10 | |
ZMXDDKWLCZADIW-UHFFFAOYSA-N | |
6228 | |
N,N-dimethylformamide |
C3H7NO | |
MFCD00003284 | |
dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa | |
CHEBI:17741 | |
CN(C)C=O |
Specifications
-61.0°C | |
White to Beige | |
100.0 | |
0.9450g/mL | |
100 mL | |
Authentic | |
Crystalline Powder | |
99.8% | |
C3H7NO | |
MFCD00003284 | |
0.8 mPa.s (20°C) | |
04, 58 | |
15, 3267 | |
Solubility in water: soluble. Other solubilities: miscible with most organic solvents | |
CN(C)C=O | |
73.10 | |
CHEBI:17741 | |
99.8% | |
N,N-Dimethylformamide, water <50 ppm |
68-12-2 | |
99.75 | |
>350°C | |
153.0°C | |
58°C | |
AcroSeal™ Glass Bottle | |
1.4290 to 1.4320 | |
0.945 | |
HCON(CH3)2 | |
Vapor Pressure: 4.9mbar at 20°C | |
0.005% Max. (Coulometric) | |
01,278; 02,153; 03,115; 04,184; 05,247; 07,124; 08,189; 09,182; 11,198; 12,203; 13,124; 14,148; 16,144 | |
dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa | |
ZMXDDKWLCZADIW-UHFFFAOYSA-N | |
N,N-dimethylformamide | |
6228 | |
73.09 | |
Extra Dry over Molecular Sieve |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Harmful in contact with skin.
Harmful if inhaled.
May damage the unborn child.
Flammable liquid and vapour.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
RUO – Research Use Only