Learn More
N-Methyl-N-(trimethylsilyl)trifluoroacetamide, 97%, Thermo Scientific Chemicals
$111.71 - $1027.73
Chemical Identifiers
CAS | 24589-78-4 |
---|---|
Molecular Formula | C6H12F3NOSi |
Molecular Weight (g/mol) | 199.25 |
MDL Number | MFCD00000411 |
InChI Key | MSPCIZMDDUQPGJ-UHFFFAOYSA-N |
Synonym | mstfa, n-methyl-n-trimethylsilyl trifluoroacetamide, 2,2,2-trifluoro-n-methyl-n-trimethylsilyl acetamide, n-methyl-n-trimethylsilyltrifluoroacetamide, acetamide, 2,2,2-trifluoro-n-methyl-n-trimethylsilyl, n-methyl-n-trimethylsilyl-trifluoroacetamide, acmc-209tfm |
PubChem CID | 32510 |
ChEBI | CHEBI:85064 |
IUPAC Name | 2,2,2-trifluoro-N-methyl-N-trimethylsilylacetamide |
SMILES | CN(C(=O)C(F)(F)F)[Si](C)(C)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1314106
|
Thermo Scientific Chemicals
A1314106 |
5 g |
Each for $111.71
|
|
|||||
AAA1314114
|
Thermo Scientific Chemicals
A1314114 |
25 g |
Each for $337.44
|
|
|||||
AAA1314122
|
Thermo Scientific Chemicals
A1314122 |
100 g |
Each for $1,027.73
|
|
|||||
Description
N-Methyl-N-(trimethylsilyl)trifluoroacetamide is used as a silylating reagent that forms volatile derivatives for GC-MS analysis. It acts as a pharmaceutical intermediate and metabolite of Famprofazone in humans. It is also used in the detection of steroid hormones 17-alfa-ethynylestradiol and estrone.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN-Methyl-N-(trimethylsilyl)trifluoroacetamide is used as a silylating reagent that forms volatile derivatives for GC-MS analysis. It acts as a pharmaceutical intermediate and metabolite of Famprofazone in humans. It is also used in the detection of steroid hormones 17-alfa-ethynylestradiol and estrone.
Solubility
Immiscible with water.
Notes
Moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents and strong acids.
Chemical Identifiers
24589-78-4 | |
199.25 | |
MSPCIZMDDUQPGJ-UHFFFAOYSA-N | |
32510 | |
2,2,2-trifluoro-N-methyl-N-trimethylsilylacetamide |
C6H12F3NOSi | |
MFCD00000411 | |
mstfa, n-methyl-n-trimethylsilyl trifluoroacetamide, 2,2,2-trifluoro-n-methyl-n-trimethylsilyl acetamide, n-methyl-n-trimethylsilyltrifluoroacetamide, acetamide, 2,2,2-trifluoro-n-methyl-n-trimethylsilyl, n-methyl-n-trimethylsilyl-trifluoroacetamide, acmc-209tfm | |
CHEBI:85064 | |
CN(C(=O)C(F)(F)F)[Si](C)(C)C |
Specifications
24589-78-4 | |
130°C to 132°C | |
C6H12F3NOSi | |
MFCD00000411 | |
UN1993 | |
Moisture sensitive | |
Immiscible with water. | |
CN(C(=O)C(F)(F)F)[Si](C)(C)C | |
199.25 | |
CHEBI:85064 | |
97% |
1.076 | |
25°C (77°F) | |
1.38 | |
5 g | |
1941550 | |
mstfa, n-methyl-n-trimethylsilyl trifluoroacetamide, 2,2,2-trifluoro-n-methyl-n-trimethylsilyl acetamide, n-methyl-n-trimethylsilyltrifluoroacetamide, acetamide, 2,2,2-trifluoro-n-methyl-n-trimethylsilyl, n-methyl-n-trimethylsilyl-trifluoroacetamide, acmc-209tfm | |
MSPCIZMDDUQPGJ-UHFFFAOYSA-N | |
2,2,2-trifluoro-N-methyl-N-trimethylsilylacetamide | |
32510 | |
199.25 | |
N-Methyl-N-(trimethylsilyl)trifluoroacetamide |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber : 246-331-6
RTECSNumber : AC9800000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only