Learn More
n-Hexane, Spectrophotometric Grade, 95+%, Thermo Scientific Chemicals
$109.18 - $323.84
Chemical Identifiers
CAS | 110-54-3 |
---|---|
Molecular Formula | C6H14 |
Molecular Weight (g/mol) | 86.18 |
MDL Number | MFCD02179311 |
InChI Key | VLKZOEOYAKHREP-UHFFFAOYSA-N |
Synonym | n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes |
PubChem CID | 8058 |
ChEBI | CHEBI:29021 |
IUPAC Name | hexane |
SMILES | CCCCCC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA32454K2
|
Thermo Scientific Chemicals
032454K2 |
1 L |
Each for $109.18
|
|
|||||
AA32454M1
|
Thermo Scientific Chemicals
032454M1 |
4 x 1 L |
Each for $323.84
|
|
|||||
AA32454K7
|
Thermo Scientific Chemicals
032454K7 |
4 L |
Each for $291.37
|
|
|||||
Description
n-Hexane used in spectrophotometry and environmental testing.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
110-54-3 | |
86.18 | |
VLKZOEOYAKHREP-UHFFFAOYSA-N | |
8058 | |
hexane |
C6H14 | |
MFCD02179311 | |
n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes | |
CHEBI:29021 | |
CCCCCC |
Specifications
-95°C | |
0.659 | |
1 L | |
Weak, Characteristic | |
1.375 | |
MFCD02179311 | |
1730733 | |
n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes | |
VLKZOEOYAKHREP-UHFFFAOYSA-N | |
hexane | |
8058 | |
86.18 | |
Spectrophotometric |
110-54-3 | |
69°C | |
−23°C (−9°F) | |
Liquid | |
C6H14 | |
UN1208 | |
14,4694 | |
Very soluble in ethanol,ethyl ether and chloroform. | |
CCCCCC | |
86.18 | |
CHEBI:29021 | |
≥95% | |
n-Hexane |
Safety and Handling
P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280i-P281-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P331-P332+P313-P363-P370+P378q-P501c
H225-H304-H315-H319-H335-H336-H361f-H372
DOTInformation : DOT Class: 3; Packing Group: II
EINECSNumber : 203-777-6
RTECSNumber : MN9275000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only