missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Butyric Acid 0.995 MP Biomedicals

Manufacturer:  MP Biochemicals LLC 10116980

Catalog No. ICN10116980



Specifications

Specifications

N-Butyric Acid
100g
99.50%
1-butyric acid
CCCC(=O)O
88.106
CHEBI:30772
3.0 (air = 1)
107-92-6
C4H8O2
FERIUCNNQQJTOY-UHFFFAOYSA-N
butanoic acid
264
88.11

Chemical Identifiers

107-92-6
88.106
1-butyric acid
CHEBI:30772
CCCC(=O)O
C4H8O2
FERIUCNNQQJTOY-UHFFFAOYSA-N
264
butanoic acid
Documents