Learn More
n-Butylethylmagnesium, 0.9M solution in heptane, AcroSeal™, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 381188000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
n-Butylethylmagnesium | |
62202-86-2,142-82-5 | |
AcroSeal™ Glass Bottle | |
CH3CH2Mg(CH2)3CH3 | |
800mL | |
magnesium, butylethyl, butylethylmagnesium, n-butylethylmagnesium, butyl ethyl magnesium, n-butyl ethyl magnesium, mvecfarlyqaunr-uhfffaoysa-n | |
YHNWUQFTJNJVNU-UHFFFAOYSA-N | |
magnesium;butane;ethane | |
112818 |
0.9M Solution in Heptane | |
0.7300g/mL | |
C6H14Mg | |
MFCD00072569 | |
0.73 | |
Solubility in water: reacts | |
C[CH2-].CCC[CH2-].[Mg+2] | |
110.48 | |
110.48 |
Chemical Identifiers
62202-86-2 | |
110.48 | |
YHNWUQFTJNJVNU-UHFFFAOYSA-N | |
112818 | |
C[CH2-].CCC[CH2-].[Mg+2] |
C6H14Mg | |
MFCD00072569 | |
magnesium, butylethyl, butylethylmagnesium, n-butylethylmagnesium, butyl ethyl magnesium, n-butyl ethyl magnesium, mvecfarlyqaunr-uhfffaoysa-n | |
magnesium;butane;ethane |
Safety and Handling
GHS H Statement
May cause drowsiness or dizziness.
Causes severe skin burns and eye damage.
May be fatal if swallowed and enters airways.
Very toxic to aquatic life with long lasting effects.
In contact with water releases
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
EINECSNumber : 263-454-
RUO â Research Use Only