Learn More
N-BOC-trans-4-Hydroxy-L-proline methyl ester, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 376250050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
N-BOC-trans-4-Hydroxy-L-proline methyl ester | |
96.0 | |
91.0°C to 96.0°C | |
Authentic | |
Glass bottle | |
MFCD00076981 | |
−65° (20°C c=1 Chloroform) | |
boc-hyp-ome, n-boc-trans-4-hydroxy-l-proline methyl ester, 2s,4r-1-tert-butyl 2-methyl 4-hydroxypyrrolidine-1,2-dicarboxylate, 1-tert-butyl 2-methyl 2s,4r-4-hydroxypyrrolidine-1,2-dicarboxylate, boc-l-hydroxyproline methyl ester, boc-hydroxyproline-ome, methyl trans-1-boc-4-hydroxy-l-prolinate, n-tert-butoxycarbonyl-trans-4-hydroxy-l-proline methyl ester, trans-1-tert-butoxycarbonyl-4-hydroxy-l-proline methyl ester, n-boc-trans-4-hydroxyl-l-proline methyl ester | |
MZMNEDXVUJLQAF-SFYZADRCSA-N | |
1-O-tert-butyl 2-O-methyl (2S,4R)-4-hydroxypyrrolidine-1,2-dicarboxylate | |
2734883 | |
97% |
74844-91-0 | |
100.0 | |
White to Beige | |
96% min. (GC) | |
C11H19NO5 | |
5 g | |
− 65.00 | |
Solubility in water: slightly soluble. | |
COC(=O)[C@@H]1C[C@@H](O)CN1C(=O)OC(C)(C)C | |
245.28 | |
245.28 | |
Crystalline Powder |
Chemical Identifiers
74844-91-0 | |
245.28 | |
MZMNEDXVUJLQAF-SFYZADRCSA-N | |
2734883 |
C11H19NO5 | |
MFCD00076981 | |
boc-hyp-ome, n-boc-trans-4-hydroxy-l-proline methyl ester, 2s,4r-1-tert-butyl 2-methyl 4-hydroxypyrrolidine-1,2-dicarboxylate, 1-tert-butyl 2-methyl 2s,4r-4-hydroxypyrrolidine-1,2-dicarboxylate, boc-l-hydroxyproline methyl ester, boc-hydroxyproline-ome, methyl trans-1-boc-4-hydroxy-l-prolinate, n-tert-butoxycarbonyl-trans-4-hydroxy-l-proline methyl ester, trans-1-tert-butoxycarbonyl-4-hydroxy-l-proline methyl ester, n-boc-trans-4-hydroxyl-l-proline methyl ester | |
COC(=O)[C@@H]1C[C@@H](O)CN1C(=O)OC(C)(C)C |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER
GHS Signal Word: Warning
RUO – Research Use Only