Learn More
N-Boc-L-pipecolinic acid, 98+%, Thermo Scientific Chemicals
$177.36 - $768.60
Chemical Identifiers
CAS | 26250-84-0 |
---|---|
Molecular Formula | C11H19NO4 |
Molecular Weight (g/mol) | 229.276 |
MDL Number | MFCD00151904 |
InChI Key | JQAOHGMPAAWWQO-QMMMGPOBSA-N |
Synonym | boc-pip-oh, s-1-boc-piperidine-2-carboxylic acid, s-n-boc-piperidine-2-carboxylic acid, s-1-tert-butoxycarbonyl piperidine-2-carboxylic acid, boc-l-pipecolic acid, boc-l-pip-oh, n-boc-l-pipecolinic acid, l-n-boc-pipecolic acid, 2s-1-tert-butoxycarbonyl piperidine-2-carboxylic acid, 2s-1-tert-butoxy carbonyl piperidine-2-carboxylic acid |
PubChem CID | 688617 |
IUPAC Name | (2S)-1-[(2-methylpropan-2-yl)oxycarbonyl]piperidine-2-carboxylic acid |
SMILES | CC(C)(C)OC(=O)N1CCCCC1C(=O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1936103
|
Thermo Scientific Chemicals
L1936103 |
1 g |
Each for $177.36
|
|
|||||
AAL1936106
|
Thermo Scientific Chemicals
L1936106 |
5 g |
Each for $768.60
|
|
|||||
Description
N-Boc-L-pipecolinic acid is a useful boc protected Pipecolic Acid for proteomics research in modulating ion channel activity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN-Boc-L-pipecolinic acid is a useful boc protected Pipecolic Acid for proteomics research in modulating ion channel activity.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
Chemical Identifiers
26250-84-0 | |
229.276 | |
JQAOHGMPAAWWQO-QMMMGPOBSA-N | |
688617 | |
CC(C)(C)OC(=O)N1CCCCC1C(=O)O |
C11H19NO4 | |
MFCD00151904 | |
boc-pip-oh, s-1-boc-piperidine-2-carboxylic acid, s-n-boc-piperidine-2-carboxylic acid, s-1-tert-butoxycarbonyl piperidine-2-carboxylic acid, boc-l-pipecolic acid, boc-l-pip-oh, n-boc-l-pipecolinic acid, l-n-boc-pipecolic acid, 2s-1-tert-butoxycarbonyl piperidine-2-carboxylic acid, 2s-1-tert-butoxy carbonyl piperidine-2-carboxylic acid | |
(2S)-1-[(2-methylpropan-2-yl)oxycarbonyl]piperidine-2-carboxylic acid |
Specifications
26250-84-0 | |
C11H19NO4 | |
1 g | |
boc-pip-oh, s-1-boc-piperidine-2-carboxylic acid, s-n-boc-piperidine-2-carboxylic acid, s-1-tert-butoxycarbonyl piperidine-2-carboxylic acid, boc-l-pipecolic acid, boc-l-pip-oh, n-boc-l-pipecolinic acid, l-n-boc-pipecolic acid, 2s-1-tert-butoxycarbonyl piperidine-2-carboxylic acid, 2s-1-tert-butoxy carbonyl piperidine-2-carboxylic acid | |
JQAOHGMPAAWWQO-QMMMGPOBSA-N | |
CC(C)(C)OC(=O)N1CCCCC1C(=O)O | |
229.276 | |
229.28 | |
N-Boc-L-pipecolinic acid |
121°C to 125°C | |
MFCD00151904 | |
3652038 | |
Insoluble in water. | |
−46° (c=1 in Methanol) | |
(2S)-1-[(2-methylpropan-2-yl)oxycarbonyl]piperidine-2-carboxylic acid | |
688617 | |
≥98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only