missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Boc-1-benzyloxymethyl-L-histidine, 97%, Thermo Scientific™
$85.68 - $85.68
Chemical Identifiers
CAS | 83468-83-1 |
---|---|
Molecular Formula | C19H25N3O5 |
Molecular Weight (g/mol) | 375.43 |
MDL Number | MFCD00038627 |
InChI Key | IEAOAWZLUGOPJX-UHFFFAOYNA-N |
Synonym | boc-l-his bom-oh, n-boc-n'-benzyloxymethyl-l-histidine, s-3-1-benzyloxy methyl-1h-imidazol-4-yl-2-tert-butoxycarbonyl amino propanoic acid, bochis bom oh, pubchem18939, n alpha-tert-butyloxycarbonyl-n pi-benzyloxyhistidine, l-histidine, n-1,1-dimethylethoxy carbonyl-1-phenylmethoxy methyl, 2s-3-1-benzyloxymethyl imidazol-4-yl-2-tert-butoxycarbonylamino propanoic acid, 2s-3-1-benzyloxy methyl imidazol-4-yl-2-tert-butoxycarbonyl amino propanoic acid |
PubChem CID | 11245813 |
IUPAC Name | (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-[1-(phenylmethoxymethyl)imidazol-4-yl]propanoic acid |
SMILES | CC(C)(C)OC(=O)NC(CC1=CN(COCC2=CC=CC=C2)C=N1)C(O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH6266303
|
Thermo Scientific Chemicals
H6266303 |
1 g |
Each for $85.68
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
83468-83-1 | |
375.43 | |
IEAOAWZLUGOPJX-UHFFFAOYNA-N | |
11245813 | |
CC(C)(C)OC(=O)NC(CC1=CN(COCC2=CC=CC=C2)C=N1)C(O)=O |
C19H25N3O5 | |
MFCD00038627 | |
boc-l-his bom-oh, n-boc-n'-benzyloxymethyl-l-histidine, s-3-1-benzyloxy methyl-1h-imidazol-4-yl-2-tert-butoxycarbonyl amino propanoic acid, bochis bom oh, pubchem18939, n alpha-tert-butyloxycarbonyl-n pi-benzyloxyhistidine, l-histidine, n-1,1-dimethylethoxy carbonyl-1-phenylmethoxy methyl, 2s-3-1-benzyloxymethyl imidazol-4-yl-2-tert-butoxycarbonylamino propanoic acid, 2s-3-1-benzyloxy methyl imidazol-4-yl-2-tert-butoxycarbonyl amino propanoic acid | |
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-[1-(phenylmethoxymethyl)imidazol-4-yl]propanoic acid |
Specifications
83468-83-1 | |
C19H25N3O5 | |
boc-l-his bom-oh, n-boc-n'-benzyloxymethyl-l-histidine, s-3-1-benzyloxy methyl-1h-imidazol-4-yl-2-tert-butoxycarbonyl amino propanoic acid, bochis bom oh, pubchem18939, n alpha-tert-butyloxycarbonyl-n pi-benzyloxyhistidine, l-histidine, n-1,1-dimethylethoxy carbonyl-1-phenylmethoxy methyl, 2s-3-1-benzyloxymethyl imidazol-4-yl-2-tert-butoxycarbonylamino propanoic acid, 2s-3-1-benzyloxy methyl imidazol-4-yl-2-tert-butoxycarbonyl amino propanoic acid | |
CC(C)(C)OC(=O)NC(CC1=CN(COCC2=CC=CC=C2)C=N1)C(O)=O | |
375.43 | |
375.42 | |
1 g | |
N-Boc-1-benzyloxymethyl-L-histidine |
White | |
MFCD00038627 | |
IEAOAWZLUGOPJX-UHFFFAOYNA-N | |
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-[1-(phenylmethoxymethyl)imidazol-4-yl]propanoic acid | |
11245813 | |
97% | |
Powder |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures