missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Acetyl-L-Tyrosine, MP Biomedicals™

$67.45 - $224.75

Chemical Identifiers

CAS 537-55-3
Molecular Formula C11H13NO4
Molecular Weight (g/mol) 223.228
InChI Key CAHKINHBCWCHCF-JTQLQIEISA-N
Synonym ac-tyr-oh
PubChem CID 68310
ChEBI CHEBI:21563
IUPAC Name (2S)-2-acetamido-3-(4-hydroxyphenyl)propanoic acid
SMILES CC(=O)NC(CC1=CC=C(C=C1)O)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN10015005 View Documents MP Biochemicals LLC
10015005
5g Each for $67.45
 
ICN10015025 View Documents MP Biochemicals LLC
10015025
25g Each for $224.75
 
Specifications

Chemical Identifiers

537-55-3
223.228
ac-tyr-oh
CHEBI:21563
CC(=O)NC(CC1=CC=C(C=C1)O)C(=O)O
C11H13NO4
CAHKINHBCWCHCF-JTQLQIEISA-N
68310
(2S)-2-acetamido-3-(4-hydroxyphenyl)propanoic acid

Specifications

537-55-3
ac-tyr-oh
CC(=O)NC(CC1=CC=C(C=C1)O)C(=O)O
223.228
CHEBI:21563
Crystalline
5g
C11H13NO4
CAHKINHBCWCHCF-JTQLQIEISA-N
(2S)-2-acetamido-3-(4-hydroxyphenyl)propanoic acid
68310
223.2
Solid
N-Acetyl-L-Tyrosine
Safety and Handling

Safety and Handling

EINECSNumber : 208-671-3

Recommended Storage : Store at 0°C.