missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-(2-Cyanoethyl)glycine, 98%, Thermo Scientific Chemicals

$56.66 - $187.49

Chemical Identifiers

CAS 3088-42-4
Molecular Formula C5H8N2O2
Molecular Weight (g/mol) 128.13
MDL Number MFCD00066903
InChI Key KZUBZCHAWPDYQX-UHFFFAOYSA-N
Synonym n-2-cyanoethyl glycine, glycine, n-2-cyanoethyl, n-2-cyanoethyl aminoacetic acid, 2-2-cyanoethyl amino acetic acid, 2-2-cyanoethylamino acetic acid, 2-cyanoethyl amino acetic acid, wln: qv1m2cn, .alpha.-.beta.'-cyanoethyl-aminoacetic acid
PubChem CID 76526
SMILES OC(=O)CNCCC#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1819906
View Documents Promotion Details
Thermo Scientific Chemicals
A1819906
5 g
Each for $56.66
 
AAA1819914
View Documents Promotion Details
Thermo Scientific Chemicals
A1819914
25 g
Each for $187.49
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

3088-42-4
128.13
KZUBZCHAWPDYQX-UHFFFAOYSA-N
76526
C5H8N2O2
MFCD00066903
n-2-cyanoethyl glycine, glycine, n-2-cyanoethyl, n-2-cyanoethyl aminoacetic acid, 2-2-cyanoethyl amino acetic acid, 2-2-cyanoethylamino acetic acid, 2-cyanoethyl amino acetic acid, wln: qv1m2cn, .alpha.-.beta.'-cyanoethyl-aminoacetic acid
OC(=O)CNCCC#N

Specifications

∼195°C (decomposition)
C5H8N2O2
UN3439
n-2-cyanoethyl glycine, glycine, n-2-cyanoethyl, n-2-cyanoethyl aminoacetic acid, 2-2-cyanoethyl amino acetic acid, 2-2-cyanoethylamino acetic acid, 2-cyanoethyl amino acetic acid, wln: qv1m2cn, .alpha.-.beta.'-cyanoethyl-aminoacetic acid
OC(=O)CNCCC#N
128.13
128.13
5 g
3088-42-4
MFCD00066903
1756230
KZUBZCHAWPDYQX-UHFFFAOYSA-N
2-(2-cyanoethylamino)acetic acid
76526
98%
N-(2-Cyanoethyl)glycine
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

EINECSNumber : 221-418-1

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only