missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-[1-(S)-Ethoxycarbonyl-3-phenylpropyl]-L-alanine-N-carboxyanhydride, 98%, Thermo Scientific™
$132.21 - $132.21
Chemical Identifiers
CAS | 84793-24-8 |
---|---|
Molecular Formula | C16H19NO5 |
Molecular Weight (g/mol) | 305.33 |
MDL Number | MFCD00272296 |
InChI Key | GFZFELCFSBCPDB-UHFFFAOYNA-N |
Synonym | s-ethyl 2-s-4-methyl-2,5-dioxooxazolidin-3-yl-4-phenylbutanoate, ethyl 2s-2-4s-4-methyl-2,5-dioxo-1,3-oxazolidin-3-yl-4-phenylbutanoate, n-1-s-+-ethoxycarbonyl-3-phenylpropyl-l-alanyl carboxyanhydride, n-1-s-ethoxycarbonyl-3-phenylpropyl-l-alanine-n-carboxyanhydride, ethyl s-2-s-4-methyl-2,5-dioxo-1,3-oxazolidin-3-yl-4-phenylbutyrate, n-1-s-ethoxycarbony1-3-phenylpropyl-l-alanine-n-carboxyanhydride, n-1-s-ethoxycarbonyl-3-phenylpropyl-n-carboxy-l-alanine anhydride nepa-nca, ecpp-aca |
PubChem CID | 9922512 |
IUPAC Name | ethyl (2S)-2-[(4S)-4-methyl-2,5-dioxo-1,3-oxazolidin-3-yl]-4-phenylbutanoate |
SMILES | CCOC(=O)C(CCC1=CC=CC=C1)N1C(C)C(=O)OC1=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1935504
|
Thermo Scientific Chemicals
L1935504 |
2 g |
Each for $132.21
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
84793-24-8 | |
305.33 | |
GFZFELCFSBCPDB-UHFFFAOYNA-N | |
9922512 | |
CCOC(=O)C(CCC1=CC=CC=C1)N1C(C)C(=O)OC1=O |
C16H19NO5 | |
MFCD00272296 | |
s-ethyl 2-s-4-methyl-2,5-dioxooxazolidin-3-yl-4-phenylbutanoate, ethyl 2s-2-4s-4-methyl-2,5-dioxo-1,3-oxazolidin-3-yl-4-phenylbutanoate, n-1-s-+-ethoxycarbonyl-3-phenylpropyl-l-alanyl carboxyanhydride, n-1-s-ethoxycarbonyl-3-phenylpropyl-l-alanine-n-carboxyanhydride, ethyl s-2-s-4-methyl-2,5-dioxo-1,3-oxazolidin-3-yl-4-phenylbutyrate, n-1-s-ethoxycarbony1-3-phenylpropyl-l-alanine-n-carboxyanhydride, n-1-s-ethoxycarbonyl-3-phenylpropyl-n-carboxy-l-alanine anhydride nepa-nca, ecpp-aca | |
ethyl (2S)-2-[(4S)-4-methyl-2,5-dioxo-1,3-oxazolidin-3-yl]-4-phenylbutanoate |
Specifications
70°C to 72°C | |
C16H19NO5 | |
5608080 | |
GFZFELCFSBCPDB-UHFFFAOYNA-N | |
ethyl (2S)-2-[(4S)-4-methyl-2,5-dioxo-1,3-oxazolidin-3-yl]-4-phenylbutanoate | |
9922512 | |
98% | |
+12° (c=2 in Acetonitrile) |
84793-24-8 | |
MFCD00272296 | |
s-ethyl 2-s-4-methyl-2,5-dioxooxazolidin-3-yl-4-phenylbutanoate, ethyl 2s-2-4s-4-methyl-2,5-dioxo-1,3-oxazolidin-3-yl-4-phenylbutanoate, n-1-s-+-ethoxycarbonyl-3-phenylpropyl-l-alanyl carboxyanhydride, n-1-s-ethoxycarbonyl-3-phenylpropyl-l-alanine-n-carboxyanhydride, ethyl s-2-s-4-methyl-2,5-dioxo-1,3-oxazolidin-3-yl-4-phenylbutyrate, n-1-s-ethoxycarbony1-3-phenylpropyl-l-alanine-n-carboxyanhydride, n-1-s-ethoxycarbonyl-3-phenylpropyl-n-carboxy-l-alanine anhydride nepa-nca, ecpp-aca | |
CCOC(=O)C(CCC1=CC=CC=C1)N1C(C)C(=O)OC1=O | |
305.33 | |
305.34 | |
2 g | |
N-[1-(S)-Ethoxycarbonyl-3-phenylpropyl]-L-alanine-N-carboxyanhydride |
Safety and Handling
GHS H Statement
H318-H317
Causes serious eye damage.
May cause an allergic skin reaction.
P261-P272-P280-P302+P352-P305+P351+P338-P310-P333+P313-P363-P501c
H317-H318
EINECSNumber : 430-360-8
TSCA : No
Recommended Storage : Ambient temperatures