missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Multi Ion cation IC standard solution, Specpure™ (Dionex) in 5% HNO{3}, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 041695AE
Description
Multi Ion cation IC standard solution is a high purity starting material, which is a certified primary reference material for analytical and Ion Chromatography.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Multi Ion cation IC standard solution | |
100 mL | |
UN3264 | |
Miscible with water |
(Dionex) in 5% HNO3 | |
MFCD03792456 | |
Specpure™ | |
Liquid |
RUO – Research Use Only