missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thiazolyl blue tetrazolium bromide, 98%, Acros Organics™

$40.81 - $236.32

Chemical Identifiers

CAS 298-93-1
Molecular Formula C18H16BrN5S
Molecular Weight (g/mol) 414.325
InChI Key AZKSAVLVSZKNRD-UHFFFAOYSA-M
Synonym thiazolyl blue, thiazolyl blue tetrazolium bromide, mmt tetrazolium, methylthiazoletetrazolium, mtt, thiazolyl blue monotetrazolium, thiazole blue, 3-4,5-dimethylthiazol-2-yl-2,5-diphenyltetrazolium bromide, mtt van, unii-euy85h477i
PubChem CID 64965
ChEBI CHEBI:53233
IUPAC Name 2-(3,5-diphenyltetrazol-2-ium-2-yl)-4,5-dimethyl-1,3-thiazole;bromide
SMILES CC1=C(SC(=N1)[N+]2=NC(=NN2C3=CC=CC=C3)C4=CC=CC=C4)C.[Br-]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC158992500 SDS View Documents Acros Organics
AC158992500
250mg Glass bottle Each for $40.81
AC158990010 SDS View Documents Acros Organics
158990010
1g Glass bottle Each for $60.25
AC158990050 SDS View Documents Acros Organics
158990050
5g Glass bottle Each for $236.32
Specifications

Chemical Identifiers

298-93-1
414.325
thiazolyl blue, thiazolyl blue tetrazolium bromide, mmt tetrazolium, methylthiazoletetrazolium, mtt, thiazolyl blue monotetrazolium, thiazole blue, 3-4,5-dimethylthiazol-2-yl-2,5-diphenyltetrazolium bromide, mtt van, unii-euy85h477i
CHEBI:53233
CC1=C(SC(=N1)[N+]2=NC(=NN2C3=CC=CC=C3)C4=CC=CC=C4)C.[Br-]
C18H16BrN5S
AZKSAVLVSZKNRD-UHFFFAOYSA-M
64965
2-(3,5-diphenyltetrazol-2-ium-2-yl)-4,5-dimethyl-1,3-thiazole;bromide

Specifications

Glass bottle
250mg
97.5
98%
thiazolyl blue, thiazolyl blue tetrazolium bromide, mmt tetrazolium, methylthiazoletetrazolium, mtt, thiazolyl blue monotetrazolium, thiazole blue, 3-4,5-dimethylthiazol-2-yl-2,5-diphenyltetrazolium bromide, mtt van, unii-euy85h477i
AZKSAVLVSZKNRD-UHFFFAOYSA-M
2-(3,5-diphenyltetrazol-2-ium-2-yl)-4,5-dimethyl-1,3-thiazole;bromide
64965
414.32
Thiazolyl blue tetrazolium bromide, 98%
195.0°C
298-93-1
100.0
C18H16BrN5S
Solubility in water: 10g/L (20°C). Other solubilities: soluble in 5% methanol, soluble in ethanol
CC1=C(SC(=N1)[N+]2=NC(=NN2C3=CC=CC=C3)C4=CC=CC=C4)C.[Br-]
414.325
CHEBI:53233
98%
Safety and Handling

Safety and Handling

Warning

EINECSNumber : 206-069-5

RTECSNumber : XF8060000

TSCA : TSCA