MP Biomedicals™ Lipoprotein-Cholesterol Concentrate
From bovine serum
Supplier: MP Biomedicals™ 0219147625
Description
A mixture of lipoprotein cholesterol, phospholipids and fatty acids. For use as a serum-free or serum-reduced media nutrient supplement, calibration of cholesterol values, and enzymatic or nonenzymatic cholesterol determinations. Offered as a low-salt solution. No preservatives. Free of enzyme inhibitors. Filtered through 0.2μm membrane filter. Cholesterol concentration: 1000-2000mg/dL.
Specifications
1000 to 2000mg/dL | |
Lipoprotein Cholesterol concentrate | |
25mL | |
0° to 5°C |
Media Nutrient Supplement, Calibration of Cholesterol Values, Enzymatic or Non-enzymatic Cholesterol Determinations | |
7.5 to 8 | |
Bovine Serum | |
Solution |