missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MP Biomedicals™ LB Medium, Large Capsules

Culture media used for the growth of bacteria.

$210.27 - $9,025.76

Specifications

Certifications/Compliance Molecular Biology Certified™
Format Capsule
For Use With (Application) For growth of bacteria
Media Format Tryptone 10g/L, Yeast extract 5g/L, NaCl 10g/L
Storage Requirements 15-30°C
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
MP113002016 View Documents MP Biomedicals™
113002016
227g Each for $210.27
 
MP113002026 View Documents MP Biomedicals™
113002026
454g Each for $331.55
 
MP113002036 View Documents MP Biomedicals™
113002036
1000g Each for $654.57
 
MP113002046 View Documents MP Biomedicals™
113002046
5000g Each for $2,909.04
 
MP113002056 View Documents MP Biomedicals™
113002056
15kg Each for $9,025.76
 
Description

Description

  • Formulations for the E. coli growth media contain 10g of tryptone and 5g of yeast extract per liter, 10g NaCl per liter
Specifications

Specifications

Molecular Biology Certified™
For growth of bacteria
15-30°C
Capsule
Tryptone 10g/L, Yeast extract 5g/L, NaCl 10g/L