missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MP Biomedicals™ ETBR Greenbag Disposal Kit
ETBR Greenbag Disposal Kit for removing Ethidium Bromide from solutions.
Supplier: MP Biomedicals™ 112350200
Description
- Each kit can remove 500 mg of Ethidium Bromide
- Each bag contains a standard amount of activated material that will safely and easily concentrate Ethidium Bromide from large liquid quantities
- The bags containing the biohazardous material can then be disposed of in a proper container
- Applications include Agarose Gel Electrophoresis, DNA and RNA purification
- Store at room temperature 15°C to 30°C
- 50 bags/kit
Specifications
ETBr Green Bag Disposal Kit | |
Single-use | |
50 Bags/Pk. |
Store at 15°C to 30°C | |
Agarose gel electrophoresis, DNA, RNA purification |
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.