missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MP Biomedicals™ Diastase
Malt
Supplier: MP Biomedicals™ 0210153980
Description
Diastase, [9000-92-4]
- Activity: 1000 Lintner u/g
- Melting Point: 175°C (dec)
Specifications
1000 U/g | |
Digests 50g of Starch in <30 min. | |
Malt | |
Diastase |
Off-White | |
4°C | |
100 g | |
Powder |