missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MP Biomedicals™ Basal Medium Eagle Vitamin Concentrate, 100X, Modified

For use with Mammalian Cell Culture

Manufacturer:  MP Biomedicals™ 1600449

 View more versions of this product

Catalog No. ICN1600449



Specifications

Specifications

5.18
BME
L-Glutamine
Sterile
100mL
-20°C
-20°C
Documents