Learn More
Molybdenum, plasma standard solution, Specpure™ Mo 1000μg/mL, Thermo Scientific Chemicals
$85.45 - $279.62
Chemical Identifiers
CAS | 7439-98-7 |
---|---|
Molecular Formula | Mo |
Molecular Weight (g/mol) | 95.95 |
MDL Number | MFCD00003465 |
InChI Key | ZOKXTWBITQBERF-UHFFFAOYSA-N |
PubChem CID | 23932 |
ChEBI | CHEBI:28685 |
IUPAC Name | molybdenum |
SMILES | [Mo] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA35758AD
|
Thermo Scientific Chemicals
035758AD |
50 mL |
Each for $85.45
|
|
|||||
AA35758AE
|
Thermo Scientific Chemicals
035758AE |
100 mL |
Each for $121.62
|
|
|||||
AA35758AP
|
Thermo Scientific Chemicals
035758AP |
500 mL |
Each for $279.62
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7439-98-7 | |
95.95 | |
ZOKXTWBITQBERF-UHFFFAOYSA-N | |
CHEBI:28685 | |
[Mo] |
Mo | |
MFCD00003465 | |
23932 | |
molybdenum |
Specifications
7439-98-7 | |
Mo | |
ZOKXTWBITQBERF-UHFFFAOYSA-N | |
molybdenum | |
23932 | |
23 hPa (17mm Hg) at 20°C | |
Molybdenum, Mo 1000μg/mL |
50 mL | |
MFCD00003465 | |
[Mo] | |
95.95 | |
CHEBI:28685 | |
Liquid |
Safety and Handling
Keep container in a well ventilated place. Keep away from combustible material. When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
H290-H314-H335
DOTInformation : DOT Class: 8; Packing Group: III
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only