Molecular Sieves, Grade 562 (Type 3A; 4-8 Mesh Beads), Fisher Chemical
Manufacturer:  Thermo Scientific M562500
Catalog No. M562-500
Specifications
Specifications
Molecular Sieves, Grade 562
4 to 8 Mesh beads
Beige
10.3 to 10.5
12736-96-8,1332-58-7,14808-60-7
Sodium Aluminosilicate, Aluminum Sodium Silicate
Pass Test
Glass Bottle
500g
Type 3A; 4-8 Mesh Beads
MFCD00131613
Solid
Documents