missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Molecular sieves 4A, powder <50 micron, Thermo Scientific Chemicals
$132.44 - $219.13
Chemical Identifiers
CAS | 70955-01-0 |
---|---|
Molecular Formula | C20H25FN2O8 |
Molecular Weight (g/mol) | 440.42 |
MDL Number | MFCD00131613 |
InChI Key | FJUZEZRZKVMAMD-UHFFFAOYNA-N |
PubChem CID | 73906282 |
IUPAC Name | (2R,3S)-2-[[(2R)-2-acetamido-3-(3-fluorophenyl)propanoyl]amino]-5-methoxy-4-methoxycarbonyl-3-methyl-5-oxopentanoic acid |
SMILES | COC(=O)C(C(C)C(NC(=O)C(CC1=CC(F)=CC=C1)NC(C)=O)C(O)=O)C(=O)OC |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC214805000
|
Thermo Scientific Chemicals
214805000 |
500 g | Plastic Bottle |
Each for $132.44
|
|
||||
AC214800010
|
Thermo Scientific Chemicals
214800010 |
1 kg | Plastic Bottle |
Each for $219.13
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Drying agentChemical Identifiers
70955-01-0 | |
440.42 | |
FJUZEZRZKVMAMD-UHFFFAOYNA-N | |
(2R,3S)-2-[[(2R)-2-acetamido-3-(3-fluorophenyl)propanoyl]amino]-5-methoxy-4-methoxycarbonyl-3-methyl-5-oxopentanoic acid |
C20H25FN2O8 | |
MFCD00131613 | |
73906282 | |
COC(=O)C(C(C)C(NC(=O)C(CC1=CC(F)=CC=C1)NC(C)=O)C(O)=O)C(=O)OC |
Specifications
18% max. | |
500 g | |
3% max. | |
Powder | |
C20H25FN2O8 | |
Solubility in water: insoluble. | |
COC(=O)C(C(C)C(NC(=O)C(CC1=CC(F)=CC=C1)NC(C)=O)C(O)=O)C(=O)OC | |
440.42 | |
Molecular sieves 4A, Powder, <50 micron |
White to Beige | |
Plastic Bottle | |
9 to 12 (10% water suspension) | |
70955-01-0 | |
MFCD00131613 | |
FJUZEZRZKVMAMD-UHFFFAOYNA-N | |
(2R,3S)-2-[[(2R)-2-acetamido-3-(3-fluorophenyl)propanoyl]amino]-5-methoxy-4-methoxycarbonyl-3-methyl-5-oxopentanoic acid | |
73906282 |
Safety and Handling
HYGROSCOPIC
RUO – Research Use Only