Minigrip™ Used IV Blood Bag Return Double Pocket Bags
Specifically designed for proper disposal of used IV bags
Description
- Features liquid-tight adhesive closure system
- Instructions for use and a biohazard logo printed on the outside pocket
- Disposable, eliminating possibility of cross contamination caused by reusing bags
- Manufactured with Micro-Tex™ polyethylene for slip-free handling
Specifications
Specimen Transport Bag | |
Biohazard symbol, liquid tight adhesive closure, score opening | |
25.4 cm | |
25.4 cm |
Specifically designed for proper disposal of used IV bags | |
10 in. | |
10 in. |