Minerals Drinking Water/Wastewater Standard (ULTRAcheck™ Blind QC Standard)
Manufacturer:  Ultra Scientific QCI710
Catalog No. US-QC-I710
Specifications
Specifications
Minerals Drinking Water/Wastewater Standard
Component concentrations in ready-to-use solution:
Alkalinity (as CaCO3)    •25-400mg/L
pH    •5-10 units Chloride    •10-250mg/L
Potassium    •2-330mg/L
Conductivity    •500-2500μmhos
Sodium    •50-300mg/L
Sulfate    •3-400mg/L
Fluoride    •1-10mg/L
Nitrate    •1-14mg/L
Traceable to NIST.
ULTRAcheck™ Blind QC Standard
Poly Bottle
500mL
Documents