Learn More
Micronova™ TMop™ Adapters
A selection of T-shaped mop adapters that work with TMop™ and Slim-T™ series slipcovers and feature a 180°swivel joint for easy surface cleaning in awkward corners, tight spaces and under equipment. Available in either electropolished stainless steel tube bar or acetal square bar (QDST).
$169.61 - $241.42
Specifications
Sterility | Non-sterile |
---|---|
Autoclavable | Autoclavable |
Resistant To | Chemicals |
Disposable | No |
Quantity | 1/Pk. |
Catalog Number | Mfr. No. | Product Type | Material | Dimensions | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Material | Dimensions | Price | Quantity | |||
19-263-614
|
Micronova™
QPT10 |
Adapter, ACME Threaded Handle Attachment | Electropolished Stainless Steel (304), Acetal | 25.4 x 2.5 cm (10 x 1 in.) |
Each for $169.61
|
|
|||
19-263-613
|
Micronova™
QPT14 |
Adapter, ACME Threaded Handle Attachment | Electropolished Stainless Steel (304), Acetal | 35.6 x 2.5 cm (14 x 1 in.) |
Each for $179.10
|
|
|||
19-166-571
|
Micronova™
QPT18 |
Adapter, ACME Threaded Handle Attachment | Stainless Steel | 45.7 x 2.5 cm (18 x 1 in.) |
Each for $218.42
|
|
|||
19-035-368
|
Micronova™
QDST10 |
Quick Disconnect Adapter | Acetal, Electropolished Stainless Steel (304), Acetal | 25.4 x 1.9 cm (10 x 0.75 in.) |
Each for $200.79
Case of 6 Each for $1,204.74
|
|
|||
19-035-369
|
Micronova™
QDST14 |
Quick Disconnect Adapter | Acetal, Electropolished Stainless Steel (304), Acetal | 35.6 x 1.9 cm (14 x 0.75 in.) |
Each for $241.42
Case of 6 Each for $1,217.64
|
|
|||
190-035-363
|
Micronova™
QDST18 |
Quick Disconnect Adapter | Acetal, Electropolished Stainless Steel (304), Acetal | 45.7 x 1.9 cm (18 x 0.75 in.) |
Each for $216.69
Case of 6 Each for $1,300.14
|
|
|||
19-035-363D
|
Micronova™
QDT10 |
Quick Disconnect Adapter | Electropolished Stainless Steel (304), Acetal | 25.4 x 2.5 cm (10 x 1 in.) |
Each for $188.95
|
|
|||
19-035-363E
|
Micronova™
QDT14 |
Quick Disconnect Adapter | Stainless Steel | 35.6 x 2.5 cm (14 x 1 in.) |
Each for $197.59
|
|
|||
19-035-363R
|
Micronova™
QDT18 |
Quick Disconnect Adapter | Electropolished Stainless Steel (304), Acetal | 45.7 x 2.5 cm (18 x 1 in.) |
Each for $216.40
|
|
|||
Description
The Micronova TMop™ Cleanroom Mop Adapters are designed to work with the TMop™ or Slim-T™ series mop covers that snap easily onto the T-shaped mop frames. The acetal square bar frame is designed for use with ‘thick sponge’ T-Mops, as it grips into the sponge. T-Mop™ frames are available with an ACME threaded end for plastic handles (QPT) or a Quick Disconnect attachment for use with SSU series stainless steel handles (QDT and QDST).
- T-shaped mop adapters with 180° swivel joint
- Easy surface cleaning in awkward corners, tight spaces and under equipment
- Works well on room floors, walls, ceilings, soft walls and equipment exteriors
- 304 Electropolished stainless-steel tube frame fits all TMop™ and Slim-T™Mop covers (QDT, QPT)
- Square acetal bar works best with the ‘thick sponge’ TMop™covers (QDST)
- QP, ACME threaded end for attachment to Micronova plastic handles
- QD, Quick Disconnect attachment to SSU series stainless handles
- Good resistance to cleaning solutions and disinfectants
- Autoclavable
Specifications
Non-sterile | |
Chemicals | |
1/Pk. |
Autoclavable | |
No | |
Store at Ambient Temperature. Rinse to remove Disinfectant before Autoclave or Storage |