missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Mezlocillin sodium salt, Thermo Scientific Chemicals
A broad spectrum antibiotic
$366.45 - $366.45
Chemical Identifiers
CAS | 59798-30-0 |
---|---|
Molecular Formula | C21H25N5NaO8S2 |
Molecular Weight (g/mol) | 562.568 |
MDL Number | MFCD07793332 |
InChI Key | SGVORSZSYKDVFT-KZHFGBRHSA-N |
PubChem CID | 123133909 |
IUPAC Name | (2R,5S,6S)-3,3-dimethyl-6-[[(2R)-2-[(3-methylsulfonyl-2-oxoimidazolidine-1-carbonyl)amino]-2-phenylacetyl]amino]-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;sodium |
SMILES | CC1(C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=CC=C3)NC(=O)N4CCN(C4=O)S(=O)(=O)C)C(=O)O)C.[Na] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ66306MC
|
Thermo Scientific Chemicals
J66306MC |
100 mg |
N/A
|
|
|||||
Description
A broad spectrum antibiotic
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
59798-30-0 | |
562.568 | |
SGVORSZSYKDVFT-KZHFGBRHSA-N | |
(2R,5S,6S)-3,3-dimethyl-6-[[(2R)-2-[(3-methylsulfonyl-2-oxoimidazolidine-1-carbonyl)amino]-2-phenylacetyl]amino]-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;sodium |
C21H25N5NaO8S2 | |
MFCD07793332 | |
123133909 | |
CC1(C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=CC=C3)NC(=O)N4CCN(C4=O)S(=O)(=O)C)C(=O)O)C.[Na] |
Specifications
Mezlocillin Sodium Salt | |
59798-30-0 | |
MFCD07793332 | |
SGVORSZSYKDVFT-KZHFGBRHSA-N | |
(2R,5S,6S)-3,3-dimethyl-6-[[(2R)-2-[(3-methylsulfonyl-2-oxoimidazolidine-1-carbonyl)amino]-2-phenylacetyl]amino]-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;sodium | |
123133909 | |
White to Yellow | |
Mezlocillin sodium salt |
Powder | |
C21H25N5NaO8S2 | |
Soluble in water or DMSO | |
CC1(C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=CC=C3)NC(=O)N4CCN(C4=O)S(=O)(=O)C)C(=O)O)C.[Na] | |
562.568 | |
561.56 | |
100 mg |
Safety and Handling
EINECSNumber : 255-640-5
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only