missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mettler Toledo™ Refracto™ 30GS Portable Refractometer

Offers an extended measuring range due to its special prism design for measurements in laboratory and production environments.

Manufacturer:  Mettler Toledo™ 51324660

Catalog No. 01-916-222


 

Description

Description

  • Compact and easy to use
  • Sapphire prism: held by a ring of hard gold plated brass. Sapphire has a higher refractive index and a better thermal conductivity than glass
  • Temperature compensation: determines the temperature and then corrects the refractive index to a standard temperature of 20°C or any other temperature the user defines
  • Plain language interface: shows results and settings in plain language
  • Ease-of-use: makes operation easy and efficient, results are displayed on backlit LCD display
  • Comprehensive result storage: stores up to 1100 results including sample identification, measurement unit, temperature compensation coefficient, date and time
  • Transfer the data whenver you want: transfer data to PC or printer using IR interface. For this purpose infrared/RS-232 converter (51325006) is required
Specifications

Specifications

±0.0005nD, ±0.2% Brix
Compact and easy to use, ideal for measurements in laboratory and production environments
0.0001nD, 0.1% Brix
250g
0 to 85% Brix
Sapphire prism
8.8 oz.
Documents