Mettler Toledo™ Excellence RM Digital Benchtop Refractometers
Benchtop refractometers allow you to determine the refractive index, BRIX, HFCS, or user-defined concentrations of a wide variety of liquids quickly and accurately. Mettler Toledo Excellence RM Digital Benchtop Refractometer has an in-built Peltier temperature control which provides both reliable temperature stability and rapid heating/cooling rates.
Specifications
Scale Range 0 to 100% Brix
Hertz 50/60Hz
Measurement Time 20 sec.
View More Specs
Products 2
Catalog Number Mfr. No. Description Temperature Range (English) Temperature Range (Metric) Refractive Index Price Quantity  
Catalog Number Mfr. No. Description Temperature Range (English) Temperature Range (Metric) Refractive Index Price Quantity  
10-376-203 View Documents Mettler Toledo™
51337003
Model RM40 41 to 212°F 5 to 100°C 1.32 to 1.7 N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
10-376-204 View Documents Mettler Toledo™
51337004
Model RM50 41 to 167°F 5 to 75°C 1.32 to 1.58 N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
  • Inert sapphire prism facilitates measurement of thin, highly volatile, dark, or viscous liquids
  • Removable top section provides ease of cleaning with simple tap water without the need for tools
  • Plug-and-play connection of accessories such as the printer, barcode scanner, LogStraight™ biometric fingerprint reader, and ErgoSens™ infrared sensor for hands-free operation
  • Assisted sample loading and clearing are possible through the addition of FillPal™ and DryPal™ units
  • Optional EQPac (Equipment Qualification Package) provides comprehensive, detailed qualification guidance (IQ/OQ/PQ) to those working in regulated environments
Expand Your Scope of Measurement
  • LiquiPhysics™ allows you to add parameters such as pH, color, and density/specific gravity to your refractive index readings
  • Simply connect a compatible SevenCompact™ pH or conductivity meter, colorimeter, or density cell to the RM module and define the method
  • Optional LabX™ LiquiPhysics™ Express software captures and manages all of these data points under a single platform
  • LabX™ also fully supports your CFR 21.11 requirements, provides user management, audit trail, flexible data export options, and fully customized reports
Reliable Standards
  • Mettler Toledo Certified Combined Density and Refractive Index Standards reduce risk, uncertainty, and error-prone sample transfer Standards come complete with certificates including an easy-to-scan barcode containing all information necessary for calibration
  • Barcodes save you valuable time, reduce data entry error, and improve traceability
Specifications
Specifications
0 to 100% Brix
50/60Hz
20 sec.