Mettler Toledo™ XPE-S Series Precision Balances
Provide outstanding weighing performance and support the highest requirements for safety, efficiency and ease of compliance
$5,338.91 - $9,974.57
Specifications
Material Solid metal housing; stainless-steel weighing pan
Electrical Requirements 100-240V AC, 0.8 A, 50/60 Hz, 12V DC 2.5 A
Connection Type RS232, Ethernet, Bluetooth, RFID
Certifications/Compliance UL Listed. CE compliant.
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Model Stabilization Time Capacity (Metric) Readability Price Quantity  
Catalog Number Mfr. No. Model Stabilization Time Capacity (Metric) Readability Price Quantity  
01-913-858 Mettler Toledo™
ME30077934
XPE204S 2 sec. 210g 0.1mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
01-913-859 Mettler Toledo™
ME30077946
XPE404S 2 sec. 410g 0.1mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
01-913-860 Mettler Toledo™
ME30114552
XPE303S 1.5 sec. 310g 1mg Each for $8,019.63
01-913-862 Mettler Toledo™
ME30115872
XPE603SDR 1.5 sec. 120 / 610g 10 / 1mg Each for $8,772.43
01-913-861 Mettler Toledo™
ME30115733
XPE603S 1.5 sec. 610g 1mg Each for $9,756.64
01-913-864 Mettler Toledo™
ME30116167
XPE1202S 1.2 sec. 1210g 10mg Each for $6,736.91
01-913-863 Mettler Toledo™
ME30115878
XPE1203S 1.5 sec. 1210g 1mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
01-913-865 Mettler Toledo™
ME30116189
XPE2002S 1.2 sec. 2.1kg 10mg Each for $7,646.84
01-913-867 Mettler Toledo™
ME30116326
XPE4001S 0.8 sec. 4.1kg 100mg Each for $5,338.91
01-913-868 Mettler Toledo™
ME30116338
XPE4002S 1.2 sec. 4.1kg 10mg Each for $8,251.04
01-913-870 Mettler Toledo™
ME30116363
XPE6001S 0.8 sec. 6.1kg 100mg Each for $6,281.27
01-913-869 Mettler Toledo™
ME30116354
XPE5003S 2 sec. 5100g 1mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
01-913-866 Mettler Toledo™
ME30116201
XPE3003S 2 sec. 3100g 1mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
01-913-872 Mettler Toledo™
ME30116397
XPE6002SDR 1.2 sec. 1.2 / 6.1kg 100 / 10mg Each for $8,658.52
01-913-871 Mettler Toledo™
ME30116387
XPE6002S 1.2 sec. 6.1kg 10mg Each for $9,078.13
01-913-873 Mettler Toledo™
ME30117876
XPE8001S 1 sec. 8.1kg 100mg Each for $7,905.73
01-913-874 Mettler Toledo™
ME30117888
XPE8002S 1.5 sec. 8.1kg 10mg Each for $9,400.06
01-913-875 Mettler Toledo™
ME30118106
XPE10002S 1.5 sec. 10.1kg 10mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
01-913-882 Mettler Toledo™
ME30122677
XPE10001S 1 sec. 10.1kg 100mg Each for $8,762.52
01-913-886 Mettler Toledo™
ME30126029
XPE6003SD5 2 sec. 6100g 5mg Each for $9,974.57
01-913-885 Mettler Toledo™
ME30126017
XPE3003SD5 2 sec. 3100g 5mg Each for $9,192.04
01-914-056 Mettler Toledo™
ME30265549
XPE603SN 1.5 sec. 610g 1mg Each for $9,115.14
01-914-055 Mettler Toledo™
ME30265517
XPE303SN 1.5 sec. 310g 1mg Each for $7,410.76
Specifications
Description
  • Durable and easy to clean
  • High resolution technology with proFACT Advanced internal adjustment (on selected models)
  • SmartPan™ weighing pan with an integrated drip tray minimizes effects of air currents on weighing cell to deliver results up to two times faster
  • Repeatability values are improved up to two-fold
  • MonoblocHighSpeed weighing cell for guaranteed performance
  • StatusLight™ indicates intuitively the status of the balance by using colors
  • LevelGuide™ provides warning when the balance is not level
  • Small footprint, ergonomic space-saver design
  • Large color touchscreen with big numbers on display
  • SmartTrac™ dosing guide
  • Compact draft shield has all-round access for handling large items
  • ErgoStand protects the display from spills and makes it easier to read
  • SmartSens infra-red sensors built into the terminal for touch-less operation
  • TestManager embedded software
  • MinWeigh function
  • FACT and GWP history
  • LabX™ software provides flexible SOP user guidance on the balance touchscreen
  • Calibry ready
  • RFID exchange with titrators
  • RFID SampleTrack
  • Built in RS232 with seven options for the second slot
Applications
Standard weighing, piece counting, percent weighing, statistics, formulation, dynamic weighing, density, differential weighing, factor calculation, RFID Exchange for titrators, RFID SampleTrack
Compliance
METTLER TOLEDO provides ISO 9001 manufacturing, nationwide ISO 9001 field service, ISO 9001 quality certification, and ISO 14001 environmental certification. UL listed. CSA certified.
Description & Specifications
Specifications
Solid metal housing; stainless-steel weighing pan
100-240V AC, 0.8 A, 50/60 Hz, 12V DC 2.5 A
RS232, Ethernet, Bluetooth, RFID
UL Listed. CE compliant.