Mettler Toledo™ Balancelink™ Software
Facilitates direct transfer of measurement results into application program
Supplier: Mettler Toledo™ 237010
Requires : MS Windows 2000™(SP4 or higher)/ MS Windows XP™ (SP1 or higher), Intel Pentium II™ 450 MHz, 128 MB RAM
Description
- Integration of highly diverse Mettler Toledo measuring instruments
- Configurable interface for integration of other instruments
- Filters and macros allow user-defined data format
- Program settings can be saved for future use
- Instrument specific configuration options can be protected and only accessed by authorised users
- Password-protected settings
- Date and time stamp for each measurement result (GLP/GMP)
- Data can be written automatically into a simple text document
- Automatic conversion of measurement values
- Supports five languages: English, French, German, Italian and Spanish
- Allows data communication between awide range of Mettler Toledo instruments and application program such as MS Word™, Excel™, MS Access™ and many more
- Online help also available
Specifications
Balance | |
Transfers data into desktop applications, integrat | |
Balancelink™ Software |