missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methylgermanium trichloride, 97%, Thermo Scientific Chemicals

$68.64 - $715.86

Chemical Identifiers

CAS 993-10-2
Molecular Formula CH9Cl3Ge
Molecular Weight (g/mol) 200.06
MDL Number MFCD00013585
InChI Key NNMJXXWNUALEKD-UHFFFAOYSA-N
Synonym methyltrichlorogermane, trichloro methyl germane, methylgermanium trichloride, germanium methyl trichloride, germane, trichloromethyl, fefxfmqvsdtspa-uhfffaoysa, inchi=1/ch3cl3ge/c1-5 2,3 4/h1h3
PubChem CID 70436
IUPAC Name trichloro(methyl)germane
SMILES Cl.Cl.Cl.C[GeH3]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4144902
View Documents Promotion Details
Thermo Scientific Chemicals
04144902
0.5 g
Each for $68.64
 
AA4144904
View Documents Promotion Details
Thermo Scientific Chemicals
04144904
2 g
Each for $207.99
 
AA4144909
View Documents Promotion Details
Thermo Scientific Chemicals
04144909
10 g
Each for $715.86
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methylgermanium trichloride is used as a precursor for the synthesis of five-coordinated spirocyclic anionic methylgermanates by reacting with catechol. It is also involved in the photo-chemical adsorption of organo-germanium on a hydrogen terminated atomically flat silicon surface using xenon excimer light.

Solubility
Miscible with ether.

Notes
Sensitive to moisture and reacts violently with water. Also hydrolysis takes place when contact with water. Incompatible with oxidizing agents and bases.
Specifications

Chemical Identifiers

993-10-2
200.06
NNMJXXWNUALEKD-UHFFFAOYSA-N
70436
Cl.Cl.Cl.C[GeH3]
CH9Cl3Ge
MFCD00013585
methyltrichlorogermane, trichloro methyl germane, methylgermanium trichloride, germanium methyl trichloride, germane, trichloromethyl, fefxfmqvsdtspa-uhfffaoysa, inchi=1/ch3cl3ge/c1-5 2,3 4/h1h3
trichloro(methyl)germane

Specifications

993-10-2
1.706 g/mL
10°C (50°F)
CH9Cl3Ge
CH3GeCl3
0.5 g
Moisture sensitive
NNMJXXWNUALEKD-UHFFFAOYSA-N
trichloro(methyl)germane
70436
97%
Methylgermanium trichloride
Colorless to Yellow
111°C
Chlorine-like
1.4685
MFCD00013585
UN2924
methyltrichlorogermane, trichloro methyl germane, methylgermanium trichloride, germanium methyl trichloride, germane, trichloromethyl, fefxfmqvsdtspa-uhfffaoysa, inchi=1/ch3cl3ge/c1-5 2,3 4/h1h3
Cl.Cl.Cl.C[GeH3]
200.06
193.99
Liquid
Safety and Handling

Safety and Handling

GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H225-H314-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.

EINECSNumber : 213-604-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only