Learn More
Methyl 4-hydroxybenzoate, 99%, Thermo Scientific Chemicals
$61.56 - $1503.20
Chemical Identifiers
CAS | 99-76-3 |
---|---|
Molecular Formula | C8H8O3 |
Molecular Weight (g/mol) | 152.15 |
MDL Number | MFCD00002352 |
InChI Key | LXCFILQKKLGQFO-UHFFFAOYSA-N |
Synonym | methylparaben, methyl paraben, methyl p-hydroxybenzoate, nipagin, methyl parahydroxybenzoate, maseptol, p-hydroxybenzoic acid methyl ester, 4-hydroxybenzoic acid methyl ester, p-carbomethoxyphenol, tegosept m |
PubChem CID | 7456 |
ChEBI | CHEBI:31835 |
IUPAC Name | methyl 4-hydroxybenzoate |
SMILES | COC(=O)C1=CC=C(C=C1)O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC126961000
|
Thermo Scientific Chemicals
126961000 |
100 g | Plastic bottle |
Each for $61.56
|
|
||||
AC126965000
|
Thermo Scientific Chemicals
126965000 |
500 g | Plastic bottle |
Each for $152.42
|
|
||||
AC126960025
|
Thermo Scientific Chemicals
126960025 |
2.5 kg | Plastic bottle |
Each for $530.92
|
|
||||
AC126960100
|
Thermo Scientific Chemicals
126960100 |
10 kg | Plastic Drum |
Each for $1,503.20
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
99-76-3 | |
152.15 | |
LXCFILQKKLGQFO-UHFFFAOYSA-N | |
7456 | |
methyl 4-hydroxybenzoate |
C8H8O3 | |
MFCD00002352 | |
methylparaben, methyl paraben, methyl p-hydroxybenzoate, nipagin, methyl parahydroxybenzoate, maseptol, p-hydroxybenzoic acid methyl ester, 4-hydroxybenzoic acid methyl ester, p-carbomethoxyphenol, tegosept m | |
CHEBI:31835 | |
COC(=O)C1=CC=C(C=C1)O |
Specifications
99-76-3 | |
White | |
0.1% max. | |
99% | |
C8H8O3 | |
MFCD00002352 | |
10,158 | |
methylparaben, methyl paraben, methyl p-hydroxybenzoate, nipagin, methyl parahydroxybenzoate, maseptol, p-hydroxybenzoic acid methyl ester, 4-hydroxybenzoic acid methyl ester, p-carbomethoxyphenol, tegosept m | |
LXCFILQKKLGQFO-UHFFFAOYSA-N | |
methyl 4-hydroxybenzoate | |
7456 | |
152.15 | |
Crystalline Powder |
125.0°C to 128.0°C | |
298.6°C | |
Authentic | |
Plastic bottle | |
HOC6H4CO2CH3 | |
100 g | |
15,618 | |
Solubility in water: 0.25% (25°C). Other solubilities: freely soluble in alcohol,acetone and ether,slightly soluble in benzene and ccl4 | |
COC(=O)C1=CC=C(C=C1)O | |
152.15 | |
CHEBI:31835 | |
99% | |
Methyl 4-hydroxybenzoate, 99% |
Safety and Handling
GHS H Statement
Harmful to aquatic life with long lasting effects.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber : 202-785-7
RTECSNumber : DH2450000
TSCA : TSCA
RUO – Research Use Only