missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methanol/Toluene Solution, 1:1 (50% Methyl Alcohol, 50% Toluene), Ricca Chemical

Manufacturer:  Ricca Chemical Company RISS0680004C

Catalog No. SS068000-4C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Methanol/Toluene Solution
53.0,48.55
methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene
Liquid
Colorless
51.0,46.65
67-56-1
1:1 (50% Methyl Alcohol, 50% Toluene)
Glass amber
4L
Documents