Learn More
Methanol, LC-MS Grade, 99.8+%, Thermo Scientific Chemicals
Solvent for LC-MS analysis
$105.73 - $831.71
Chemical Identifiers
CAS | 67-56-1 |
---|---|
Molecular Formula | CH4O |
Molecular Weight (g/mol) | 32.04 |
MDL Number | MFCD00004595 |
InChI Key | OKKJLVBELUTLKV-UHFFFAOYSA-N |
Synonym | methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit |
PubChem CID | 887 |
ChEBI | CHEBI:17790 |
IUPAC Name | methanol |
SMILES | CO |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA47192K2
|
Thermo Scientific Chemicals
047192K2 |
1 L |
Each for $105.73
|
|
|||||
AA47192M1
|
Thermo Scientific Chemicals
047192M1 |
4 x 1 L |
Pack of 4 Each for $378.42
|
|
|||||
AA47192K7
|
Thermo Scientific Chemicals
047192K7 |
4 L |
Each for $252.21
|
|
|||||
AA47192M6
|
Thermo Scientific Chemicals
047192M6 |
4 x 4 L |
Pack for $831.71
|
|
|||||
Description
Solvent for LC-MS analysisMethanol is used as a solvent in Liquid chromatography-mass spectrometry
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
67-56-1 | |
32.04 | |
OKKJLVBELUTLKV-UHFFFAOYSA-N | |
887 | |
methanol |
CH4O | |
MFCD00004595 | |
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit | |
CHEBI:17790 | |
CO |
Specifications
-98°C | |
Colorless | |
64.7°C | |
11°C (52°F) | |
Liquid | |
CH4O | |
MFCD00004595 | |
1098229 | |
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit | |
OKKJLVBELUTLKV-UHFFFAOYSA-N | |
methanol | |
887 | |
32.04 | |
LC-MS |
67-56-1 | |
0.791 g/mL | |
1 L | |
Mild | |
1.329 | |
CH3OH | |
UN1230 | |
14,5957 | |
Miscible with water and most of the organic solvents including ethyl acetate,toluene,dichloromethane,chloroform,tetrahydrofuran,and acetone. | |
CO | |
32.04 | |
CHEBI:17790 | |
≥99.8% | |
Methanol |
Safety and Handling
GHS H Statement
H225-H301-H311-H331-H370
Highly flammable liquid and vapor.
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P307+P311-P311-P312-P330-P363-P370+P378q-P501c
H225-H301+H311+H331-H370-H372
DOTInformation : Transport Hazard Class: 3; Packaging Group: II; Proper Shipping Name: METHANOL
EINECSNumber : 200-659-6
RTECSNumber : PC1400000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only