missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methanol-d4, for NMR, 99.8% atom D, ACROS Organics™

$111.35 - $389.59

Chemical Identifiers

CAS 811-98-3
Molecular Formula CH4O
Molecular Weight (g/mol) 36.066
MDL Number MFCD00044637
InChI Key OKKJLVBELUTLKV-MZCSYVLQSA-N
Synonym methanol-d4, perdeuteromethanol, tetradeuteromethanol, methan-d3-ol-d, 2h4 methanol, methyl alcohol-d4, cd3od, methyl-d3 alcohol-d, trideuterio deuteriooxy methane, methanol-d4, 99.8 atom % d
PubChem CID 71568
IUPAC Name trideuterio(deuteriooxy)methane
SMILES CO
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC321280050 SDS View Documents Acros Organics
321280050
5g Glass bottle Each for $111.35
AC321280100 SDS View Documents Acros Organics
321280100
10mL Glass bottle Each for $171.80
AC321280250 SDS View Documents Acros Organics
321280250
25mL Glass bottle Each for $389.59
Description

Description

For NMR
Specifications

Chemical Identifiers

811-98-3
36.066
OKKJLVBELUTLKV-MZCSYVLQSA-N
71568
CO
CH4O
MFCD00044637
methanol-d4, perdeuteromethanol, tetradeuteromethanol, methan-d3-ol-d, 2h4 methanol, methyl alcohol-d4, cd3od, methyl-d3 alcohol-d, trideuterio deuteriooxy methane, methanol-d4, 99.8 atom % d
trideuterio(deuteriooxy)methane

Specifications

0.8880g/mL
Glass bottle
Colorless
811-98-3
-99.0°C
CD3OD
MFCD00044637
01, III, 1187
OKKJLVBELUTLKV-MZCSYVLQSA-N
trideuterio(deuteriooxy)methane
71568
Liquid
Methanol-d4, 99.8 atom%D
99.75% min.
0.05% max. (Coulometric)
5g
65.0°C
CH4O
12°C
1.3262
methanol-d4, perdeuteromethanol, tetradeuteromethanol, methan-d3-ol-d, 2h4 methanol, methyl alcohol-d4, cd3od, methyl-d3 alcohol-d, trideuterio deuteriooxy methane, methanol-d4, 99.8 atom % d
CO
36.066
36.06
NMR
Safety and Handling

Safety and Handling

GHS H Statement:
Causes damage to organs.
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Highly flammable liquid and vapor.

GHS P Statement:
Immediately call a POISON CENTER or doctor/physician.
IF exposed: Call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger