missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methanol, Anhydrous, 99%, Alfa Aesar

$37.09 - $65.38

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.042
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA41467AK SDS View Documents Alfa Aesar
41467AK
250mL Each for $37.09
AA41467K2 SDS View Documents Alfa Aesar
41467K2
1L Each for $65.38
Specifications

Chemical Identifiers

67-56-1
32.042
OKKJLVBELUTLKV-UHFFFAOYSA-N
887
methanol
CH4O
MFCD00004595
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
CHEBI:17790
CO

Specifications

67-56-1
CH4O
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
CO
32.042
CHEBI:17790
Liquid
250mL
99.9%
MFCD00004595
OKKJLVBELUTLKV-UHFFFAOYSA-N
methanol
887
32.04
0.791g/mL
Methanol, anhydrous
Safety and Handling

Safety and Handling

EINECSNumber : 200-659-6