missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Specifications
Methanol | |
67-56-1 | |
0.79g/cm3 at 20°C | |
Closed cup: 12°C (53.6°F), Open cup: 15.85°C (60.5°F) | |
Alcohol-like | |
Liquid | |
CH3OH | |
UN 1230 | |
Soluble in water | |
CO | |
32.04 | |
CHEBI:17790 | |
≥99.9% |
-97.8°C | |
Colorless | |
1 L | |
464°C (867.2°F) | |
Poly Bottle | |
CH4O | |
MFCD00004595 | |
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit | |
OKKJLVBELUTLKV-UHFFFAOYSA-N | |
methanol | |
887 | |
32.04 | |
ACS/NF/USP |
Chemical Identifiers
67-56-1 | |
32.04 | |
OKKJLVBELUTLKV-UHFFFAOYSA-N | |
887 | |
methanol |
CH4O | |
MFCD00004595 | |
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit | |
CHEBI:17790 | |
CO |
Safety and Handling
DOTInformation : DOT Class: 3, Packaging Group: II
Recommended Storage : Room Temperature