missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methanol, 99+%, extra pure, ACROS Organics™

$42.25 - $383.92

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.042
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym Methyl alcohol
PubChem CID 887
ChEBI CHEBI:17790
View More Specs

Products 4 Specifications

Chemical Identifiers

67-56-1
32.042
OKKJLVBELUTLKV-UHFFFAOYSA-N
887
CH4O
MFCD00004595
Methyl alcohol
CHEBI:17790

Specifications

67-56-1
99+%
CH3OH
1230
11,322; 13,337
Methyl alcohol
OKKJLVBELUTLKV-UHFFFAOYSA-N
methanol
887
32.04
0.7910g/mL
1.3280 to 1.3300
0.791
12°C (53.6°F)
64.7°C
-98.0°C
Methanol
Extra Pure
Vapor Pressure: 128hPa at 20°C
CH4O
MFCD00004595
01, IV, 1227
15, 6029
Solubility in water: miscible with water. Other solubilities: miscible with ethanol, ether, benzene, ketones and, most other organic solvents
CO
32.042
CHEBI:17790
Liquid
Glass Bottle
0.005% max.
0.2% max. (K.F.)
0.55 mPa.s (20°C)
Undesignated
1L
≥99%
Safety and Handling

Safety and Handling

GHS H Statement:
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.
Highly flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger