missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methanol, 99.9%, for analysis, ACROS Organics™

$29.52 - $79.88

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.042
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym Methyl alcohol
PubChem CID 887
ChEBI CHEBI:17790
View More Specs

Products 5 Description

Description

For analysis
Specifications

Chemical Identifiers

67-56-1
32.042
OKKJLVBELUTLKV-UHFFFAOYSA-N
887
CH4O
MFCD00004595
Methyl alcohol
CHEBI:17790

Specifications

67-56-1
99.9%
CH3OH
Methyl alcohol
OKKJLVBELUTLKV-UHFFFAOYSA-N
50ppb max.
32.042
CHEBI:17790
Liquid
50ppb max.
50ppb max.
50ppb max.
Plastic drum
0.002% max.
0.791
0.0003 meq/g max.
50ppb max.
12°C
64.7°C
-98.0°C
Methanol
For analysis
0.001% max.
CH4O
MFCD00004595
Solubility in water: miscible with water. Other solubilities: miscible with ethanol, ether, benzene, ketones and, most other organic solvents
CO
methanol
887
32.04
50ppb max.
0.7910g/mL
50ppb max.
50ppb max.
1.3280 to 1.3300
Solubility in water: miscible with water. Other solubilities: miscible with ethanol, ether, benzene, ketones and, most other organic solvents
Passes Test
0.0002 meq/g max.
0.05% max. (K.F.)
0.55 mPa.s (20°C)
Undesignated
10L
99.9%
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.
Highly flammable liquid and vapor.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

Danger