missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methanol, 99.8%, for spectroscopy ACS, ACROS Organics™

$45.48 - $45.48

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.042
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC413770025 SDS View Documents Acros Organics
AC413770025
2.5L Glass bottle N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC413775000 SDS View Documents Acros Organics
413775000
500mL EcoSafPak™ Glass Bottle Each for $45.48
Description

Description

For UV/VIS spectroscopy
Specifications

Chemical Identifiers

67-56-1
32.042
OKKJLVBELUTLKV-UHFFFAOYSA-N
887
methanol
CH4O
MFCD00004595
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
CHEBI:17790
CO

Specifications

0.01 max. at 280nm, 0.01 max. at 400nm, 0.04 max. at 260nm, 0.1 max. at 240nm, 0.2 max. at 230nm, 0.4 max. at 220nm, 0.8 max. at 210nm, 1 max. at 205nm
Vapor Pressure: 128hPa at 20°C
CH4O
MFCD00004595
11,322; 13,337
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
OKKJLVBELUTLKV-UHFFFAOYSA-N
methanol
887
32.04
0.001% max. (each of acetone, formaldehyde, acetaldehyde)
ACS
Glass bottle
0.001% max.
0.791
Passes Test
0.0002 meq/g max.
12°C
64.7°C
-98.0°C
Methanol
67-56-1
99.8%
CH3OH
01, IV, 1227
15, 6029
Solubility in water: miscible with water. Other solubilities: miscible with ethanol, ether, benzene, ketones and, most other organic solvents
CO
32.042
CHEBI:17790
Liquid
99.8%
0.7910g/mL
1.3280 to 1.3300
Solubility in water: miscible with water. Other solubilities: miscible with ethanol, ether, benzene, ketones and, most other organic solvents
Passes Test
0.0003 meq/g max.
0.1% max. (K.F.)
0.55 mPa.s (20°C)
Undesignated
2.5L
Safety and Handling

Safety and Handling

GHS H Statement:
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.
Highly flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 200-659-6

RTECSNumber : PC1400000

TSCA : TSCA