Learn More
Methallyl phenyl ether, 96%, Thermo Scientific Chemicals
$88.67 - $330.75
Chemical Identifiers
CAS | 5820-22-4 |
---|---|
Molecular Formula | C10H12O |
Molecular Weight (g/mol) | 148.21 |
MDL Number | MFCD00008597 |
InChI Key | LECDNXOCIPRJNJ-UHFFFAOYSA-N |
Synonym | methallyl phenyl ether, 2-methylallyloxy benzene, ether, 2-methylallyl phenyl, benzene, 2-methyl-2-propenyl oxy, 2-methylallyl phenyl ether, 2-methylprop-2-en-1-yl oxy benzene, 2-methyl-2-propen-1-yl oxy benzene, phenyl-methallyl ether, acmc-1b20w |
PubChem CID | 79893 |
IUPAC Name | 2-methylprop-2-enoxybenzene |
SMILES | CC(=C)COC1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2236906
|
Thermo Scientific Chemicals
B2236906 |
5 g |
Each for $88.67
|
|
|||||
AAB2236914
|
Thermo Scientific Chemicals
B2236914 |
25 g |
Each for $330.75
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
5820-22-4 | |
148.21 | |
LECDNXOCIPRJNJ-UHFFFAOYSA-N | |
79893 | |
CC(=C)COC1=CC=CC=C1 |
C10H12O | |
MFCD00008597 | |
methallyl phenyl ether, 2-methylallyloxy benzene, ether, 2-methylallyl phenyl, benzene, 2-methyl-2-propenyl oxy, 2-methylallyl phenyl ether, 2-methylprop-2-en-1-yl oxy benzene, 2-methyl-2-propen-1-yl oxy benzene, phenyl-methallyl ether, acmc-1b20w | |
2-methylprop-2-enoxybenzene |
Specifications
5820-22-4 | |
204°C | |
C10H12O | |
MFCD00008597 | |
1933868 | |
LECDNXOCIPRJNJ-UHFFFAOYSA-N | |
2-methylprop-2-enoxybenzene | |
79893 | |
96% |
0.964 | |
77°C (171°F) | |
1.515 | |
5 g | |
methallyl phenyl ether, 2-methylallyloxy benzene, ether, 2-methylallyl phenyl, benzene, 2-methyl-2-propenyl oxy, 2-methylallyl phenyl ether, 2-methylprop-2-en-1-yl oxy benzene, 2-methyl-2-propen-1-yl oxy benzene, phenyl-methallyl ether, acmc-1b20w | |
CC(=C)COC1=CC=CC=C1 | |
148.21 | |
148.21 | |
Methallyl phenyl ether |
Safety and Handling
GHS H Statement
H334-H227-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Combustible liquid.
May cause an allergic skin reaction.
P210-P261-P272-P280-P285-P302+P352-P304+P341-P333+P313-P342+P311-P363-P370+P378q-P501c
H227-H317-H334
EINECSNumber : 227-393-3
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only