missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific Chemicals MES, 0.2M buffer soln., pH 6.0, low endotoxin
$127.60 - $207.99
Chemical Identifiers
CAS | 145224-94-8 |
---|---|
Molecular Formula | C6H15NO5S |
Molecular Weight (g/mol) | 213.248 |
MDL Number | MFCD00283848 |
InChI Key | MIIIXQJBDGSIKL-UHFFFAOYSA-N |
PubChem CID | 16218417 |
IUPAC Name | 2-morpholin-4-ylethanesulfonic acid;hydrate |
SMILES | C1COCCN1CCS(=O)(=O)O.O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ67264AE
|
Thermo Scientific Chemicals
J67264AE |
100 mL |
N/A
|
|
|||||
AAJ67264AK
|
Thermo Scientific Chemicals
J67264AK |
250 mL |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
145224-94-8 | |
213.248 | |
MIIIXQJBDGSIKL-UHFFFAOYSA-N | |
2-morpholin-4-ylethanesulfonic acid;hydrate |
C6H15NO5S | |
MFCD00283848 | |
16218417 | |
C1COCCN1CCS(=O)(=O)O.O |
Specifications
145224-94-8 | |
0.2 M | |
Liquid | |
C6H15NO5S | |
MIIIXQJBDGSIKL-UHFFFAOYSA-N | |
2-morpholin-4-ylethanesulfonic acid;hydrate | |
16218417 |
Colorless | |
6.0 | |
100 mL | |
MFCD00283848 | |
C1COCCN1CCS(=O)(=O)O.O | |
213.248 | |
MES, low endotoxin |
Safety and Handling
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only