missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Medegen Biohazardous Waste Bags, LLDPE Film, Coreless Roll
These Biohazardous Waste Bags pass ASTMD1709-85, 165 Gram Dart Test.
Supplier: Medegen 117M
Description
- Made from low-density polyethylene
- Minimize the risk of contamination from biohazardous materials during collection and transport
- Bags are red with ''Biohazardous Waste/Infectious Waste'' and biohazard symbol imprinted in black ink
- Individually folded
Specifications
Biohazardous Waste Bag | |
113.6 L | |
Red, Black | |
Biohazardous or infectious waste with biohazard symbol | |
0.0304 mm | |
30.5 x 41 in. (77.47 x 104.14cm) | |
77.47 cm | |
104.14 cm |
30 gal. | |
ASTM Dart | |
Used to collect, store and transport biohazardous and infectious waste within the health care facility, outpatient surgery center, alternate care clinic, physician's office and lab | |
LLDPE | |
1.2 mil | |
30.5 in. | |
41 in. |
May NOT be used in CA, FL, PA or for interstate (between state) transport.