Thermo Scientific™ Manual GC Syringes
Easily accomplish accurate, precise, and reproducible manual injections with Thermo Scientific™ GC Syringes.
$64.20 - $333.65
Specifications
Disposable No
For Use With (Equipment) GC Autosampler
View More Specs
Products 7
Catalog Number Mfr. No. Type Volume (Metric) Length (Metric) Price Quantity  
Catalog Number Mfr. No. Type Volume (Metric) Length (Metric) Price Quantity  
35-007-104774 View Documents Thermo Scientific™
365C0741
Fixed 5µL 50mm Each for $96.68
36-5D1-091 View Documents Thermo Scientific™
365D1091
Fixed 10µL 50mm Each for $64.20
36-5D1-096 View Documents Thermo Scientific™
365D1096
Fixed 10µL 50mm Each for $333.65
36-5F1-891 View Documents Thermo Scientific™
365F1891
Fixed 25µL 50mm Each for $97.24
36-5H2-291 View Documents Thermo Scientific™
365H2291
Manual Fixed 100µL 50mm Each for $109.61
36-5I2-541 View Documents Thermo Scientific™
365I2541
Fixed 250µL 50mm Each for $119.52
36-5G2-091 View Documents Thermo Scientific™
365G2091
Removable 50µL 50mm Each for $128.31
Description
Description
Achieve accurate, precise, and reproducible manual injections easily using Thermo Scientific™ Manual GC Syringes. The robust, field-proven designs are optimized for septum piercing. They are made of borosilicate glass with metal flanges of either stainless steel or nickel-plated brass and cover a range of volumes. Easy-to-read markings ensure reproducibility.
  • Syringes available in a range of volumes
  • PTFE Luer-Lok™ syringes
  • PTFE Luer Slip syringes
Specifications
Specifications
No
GC Autosampler
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.