missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Manganese powder, -325 mesh, 99.3% (metals basis), Thermo Scientific Chemicals
Form: Apparent density 2.6-3.5g/cm3
$46.30 - $259.12
Chemical Identifiers
CAS | 7439-96-5 |
---|---|
Molecular Formula | Mn |
Molecular Weight (g/mol) | 54.94 |
MDL Number | MFCD00011111 |
InChI Key | PWHULOQIROXLJO-UHFFFAOYSA-N |
Synonym | colloidal, mangan, manganese, elemental, cutaval, magnacat, metal alloy, tronamang, fume, mangan polish, compounds |
PubChem CID | 23930 |
ChEBI | CHEBI:35154 |
IUPAC Name | manganese |
SMILES | [Mn] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA1023814
|
Thermo Scientific Chemicals
01023814 |
25 g |
Each for $46.30
|
|
|||||
AA1023830
|
Thermo Scientific Chemicals
01023830 |
250 g |
Each for $100.42
|
|
|||||
AA10238A1
|
Thermo Scientific Chemicals
010238A1 |
1 kg |
Each for $259.12
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7439-96-5 | |
54.94 | |
PWHULOQIROXLJO-UHFFFAOYSA-N | |
23930 | |
manganese |
Mn | |
MFCD00011111 | |
colloidal, mangan, manganese, elemental, cutaval, magnacat, metal alloy, tronamang, fume, mangan polish, compounds | |
CHEBI:35154 | |
[Mn] |
Specifications
Gray | |
Gray powder | |
Mn | |
25 g | |
PWHULOQIROXLJO-UHFFFAOYSA-N | |
manganese | |
23930 | |
-325 Mesh | |
Manganese |
7439-96-5 | |
99.3% (Metals basis) | |
MFCD00011111 | |
colloidal, mangan, manganese, elemental, cutaval, magnacat, metal alloy, tronamang, fume, mangan polish, compounds | |
[Mn] | |
54.94 | |
CHEBI:35154 | |
Powder |
Safety and Handling
P210-P240-P241-P264b-P280-P305+P351+P338-P370+P378q
H228-H319
DOTInformation : DOT Class: 4.1, Packaging Group: III
EINECSNumber : 231-105-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only